Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VEGF Receptor 3 Monoclonal Antibody (AFL4), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5013415
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-134-15 100 μg
50-134-14 50 μg
50-134-16 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-134-15 Supplier Invitrogen™ Supplier No. 14598882
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The AFL4 monoclonal antibody reacts with the mouse VEGF receptor-3, also known as Flt-4. This 195 kDa molecule was identified as an endothelial-specific member of the receptor tyrosine kinase (RTK) family. During early embryogenesis all endothelial cells express VEGFR-3, while in the adult tissues, VEGFR-3 expression disappears from the vascular endothelial cells and is observed only on the lymphatic endothelium. However, VEGFR-3 expression is induced in the adult tissue upon tumor implementation suggesting an important role for this receptor in the tumor angiogenesis. VEGF-C and VEGF-D bind to and activate VEGFR-3. AFL4 is an antagonist mAb. Applications Reported: The AFL4 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting (WB), and immunohistochemical staining of frozen tissue sections. It has also been reported in blocking of ligand binding. (Please use Functional Grade purified AFL4, cat. 16-5988, in functional assays.). Applications Tested: The AFL4 antibody has been tested by flow cytometric analysis of in vitro differentiated mouse endothelial cells. In brief, mouse ES cells were incubated on collagen IV matrix for 4 days and subsequently stimulated with VEGF under serum free conditions to induce further differentiation. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE.

Vascular endothelial growth factor is a key regulator of blood vessel development in embryos and angiogenesis in adult tissues. Unlike VEGF, the related VEGFC stimulates the growth of lymphatic vessels through its specificl ymphatic endothelial receptor VEGFR-3. In 1998, Dumont showed that targeted inactivation of the VEGFR-3 gene in mice resulted in defective blood vessel development in early embryos. Vasculogenesis and angiogenesis occurred, but large vessels became abnormally organized with defective lumens, leading to fluid accumulation in the pericardial cavity and cardiovascular failure at embryonic day 9.5. Thus, VEGFR3 has an essential role in the development of theembryonic cardiovascular system before the emergence of the lymphatic vessels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF Receptor 3
Applications Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation, Neutralization, Western Blot
Classification Monoclonal
Clone AFL4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene FLT4
Gene Accession No. P35917
Gene Alias AI323512; Chy; chylous ascites; Flt4; FLT-4; FLT41; fms related tyrosine kinase 4; FMS-like tyrosine kinase 4; fms-related tyrosine kinase 4; LMPH1A; PCL; receptor protein tyrosine kinase; soluble VEGFR 3; sVEGFR 3; tyrosine-protein kinase receptor FLT4; Vascular endothelial growth factor receptor 3; vascular endothelial growth factor receptor-3; VEGF R3; VEGFR3; VEGFR-3
Gene Symbols FLT4
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14257
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.