Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ VEGFR3/Flt-4 Antibody Pair [HRP]
SDP

Catalog No. NB009635 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009635 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009635 Supplier Novus Biologicals™ Supplier No. NBP27929815PLATES
Add to Cart
Add to Cart

Mouse Monoclonal Antibody Pair

specifically detects in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 156.1 pg/mL
Assay Sensitivity 78.13-5000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human VEGFR3/Flt-4.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias EC 2.7.10, EC 2.7.10.1, FLT-4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4
Gene ID (Entrez) 2324
Gene Symbol FLT4
Host Species Mouse
Kit Contents
  • Standard: Each vial contains 80 ng of recombinant VEGFR3/Flt-4.
  • Capture Antibody: 0.5 mg/mL of mouse anti-VEGFR3/Flt-4 monoclonal antibody.
  • Detection Antibody: 0.4 mg/mL mouse anti-VEGFR3/Flt-4 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Angiogenesis, Cancer, Hypoxia, Protein Kinase
    Storage Requirements Storage is content dependent.
    Target VEGFR3/Flt-4
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating 2. Detection Antibody: Dilute to working concentration of 1 μg/mL in detection antibody dilution buffer before use 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A six-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 5 ng/mL is recommended
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.