Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI DECON-HAND™ Hand Sanitizer

Catalog No. 19546775 Shop All VAI Products
Change view
Click to view available options
Volume (English):
16 oz.
32 oz.
Sterility:
Non-sterile
Sterile
Container Type:
Bottle
Bottle for ASEPTI-CLEANSE Dispenser
Bottle with Attachable Pump
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Sterility Container Type
19-546-775 32 oz. Non-sterile Bottle for ASEPTI-CLEANSE Dispenser
19-546-838 16 oz. Non-sterile Bottle with Attachable Pump
19-546-839 16 oz. Sterile Bottle with Attachable Pump
19-546-840 32 oz. Sterile Bottle
19-546-842 16 oz. Non-sterile -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-546-775 Supplier VAI Supplier No. DH09
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Lot-specific sterile documentation

Gelled alcohol DECON-HAND™ Hand Sanitizer decreases bacteria and fungi on skin.

  • An instant hand sanitizer before glove donning
  • Apply to hands and allow to dry without wiping
  • Provides effective sanitization against bacteria and fungi according to USP 25 criteria
  • Validated for sterility and shelf life

Compliance

Manufactured in accordance with 21 CFR Part 211 Good Manufacturing Practices for DRUGS and the Tentative Final Monograph for Topical Antimicrobial Drug Products for Over-the-Counter human use.

Specifications

Product Type Hand Sanitizer
Form Gel
Sterility Non-sterile
Container Type Bottle for ASEPTI-CLEANSE Dispenser
For Use With (Application) For use as an instant hand sterilizer before glove donning
For Use With (Equipment) ASEPTI-CLEANSE Dispenser
Packaging Bottle
Volume (English) 32 oz.
Volume (Metric) 946.35 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.