Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI™ Core2Clean System™ Spare Parts

Catalog No. 19546805 Shop All VAI Products
Change view
Click to view available options
Product Type:
Clamp
Pressure Relief Valve (Automatic)
Pressure Relief Valve (Manual)
Regulator Assembly (Replacement Input)
Regulator Assembly (Replacement Output)
Source Pressure Hose
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-546-805 Regulator Assembly (Replacement Input)
19-546-806 Regulator Assembly (Replacement Output)
19-546-807 Pressure Relief Valve (Manual)
19-546-808 Pressure Relief Valve (Automatic)
19-546-799 Source Pressure Hose
19-546-797 Clamp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-546-805 Supplier VAI™ Supplier No. C2C2001
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Core2Clean™ Plus Systems were created by Veltek Associates, Inc., to provide specialized cleaning equipment for use in controlled environments.

  • Regulator, complete unit and fittings for regulating Core2Clean incoming air pressure
  • Pressure Relief Valve Assembly, automatic safety relief valve, factory set to prevent over pressurization of Core2Clean tank
  • Stainless Steel

Specifications

Product Type Regulator Assembly (Replacement Input)
For Use With (Equipment) Core2Clean Plus Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.