Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Venezuelan Equine Encephalitis Virus NSP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144448
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144448 Supplier Invitrogen™ Supplier No. PA5144448
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Venezuelan Equine Encephalitis Virus NSP1 Polyclonal Antibody for Western Blot, ELISA

Venezuelan equine encephalitis virus (VEEV) is a member of genus Alphavirus in the family Togaviridae VEEV encodes four nonstructural proteins (nsP1, nsP2, nsP3 and nsP4) and five structural proteins (capsid (C), envelope (E) 3, E2, 6k, and E1) The Nsp1 gene of VEEV contains a 51-nt conserved sequence element (CSE) that is essential for virus replication Mutation or deletion of CSE results in loss of infectivity of VEEV, probably due to inefficient recognition of the viral RNA core promoter element by the virus replication complex Nsp1 functions as a methyltransferase and guanylylation enzyme, two properties needed for the 5' capping of VEEV genomic RNA [PMID: 30781656].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Venezuelan Equine Encephalitis Virus NSP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.912 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Alias VEEV NSP1
Host Species Rabbit
Immunogen Recombinant Venezuelan equine encephalitis virus (strain Trinidad donkey) (VEEV) Non-structural polyprotein (27-167aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.