Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Venezuelan Equine Encephalitis Virus NSP2 Recombinant Rabbit Monoclonal Antibody (HL1919)
GREENER_CHOICE

Catalog No. FEMA550347 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550347 Supplier Invitrogen™ Supplier No. MA550347
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Venezuelan Equine Encephalitis Virus NSP2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

The Venezuelan Equine Encephalitis Virus NSP2 is a gene target of the Venezuelan equine encephalitis virus (VEEV), which belongs to the Togaviridae family of viruses. The NSP2 gene encodes for a non-structural protein that plays an important role in the replication and pathogenesis of the virus. VEEV is transmitted to humans and animals through the bites of infected mosquitoes, and can cause severe symptoms such as fever, headache, and encephalitis. The NSP2 gene is an important target for the development of antiviral drugs and vaccines against VEEV, as it is essential for the virus to replicate and spread in the host.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Venezuelan Equine Encephalitis Virus NSP2
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1919
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias VEEV NSP2
Host Species Rabbit
Immunogen Recombinant fragment of Venezuelan Equine Encephalitis Virus nsP2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.