Learn More
Market Lab Venipuncture Arm Warmer, Medium, 7""""L x 6""""W, 10/pk
Supplier:  Market Lab 7891
Encompass_Preferred
Simply flex the stainless steel disc to activate. Reaches 44°C (111.2°F) in under a minute. Flexible, laminated nylon molds easily to patient's arm. Non-toxic.
Catalog No. NC1154152
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.