Learn More
Selleck Chemical LLC Venlafaxine HCl 25mg 99300-78-4
SDP
Supplier:  Selleck Chemical LLC S144125MG
Encompass_Preferred
Venlafaxine HCl is an arylalkanolamine serotonin-norepinephrine reuptake inhibitor (SNRI), used to treat major depressive disorder (MDD), generalised anxiety disorder (GAD), panic disorder and social phobia. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-633-3