Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Venlafaxine Hydrochloride, Serotonin-norepinephrine Reuptake Inhibitor, Solid, 10 mg

Catalog No. 502707345
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7345 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7345 Supplier Abcam Supplier No. AB12071510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent antidepressant serotonin-norepinephrine reuptake inhibitor with greater than 99% purity.

  • Molecular Weight: 313.87 g/mol
  • Purity: >99%
  • Form: Solid
  • Activity: Serotonin-norepinephrine reuptake inhibitor (antidepressant)
  • In vitro properties: Ki values of 82 nM for SERT and 2480 nM for NET; displays 30-fold higher affinity for SERT than NET

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Serotonin Transporter, Norepinephrine Transporter
Form Solid
Content And Storage Store at ambient temperature.
Shipping Condition Ambient
Formulation 100% Venlafaxine hydrochloride
Quantity 10 mg
For Use With (Application) Antidepressant
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.