Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Venlafaxine Hydrochloride, Serotonin-Norepinephrine Reuptake Inhibitor, Solid, 99% Purity, 50 mg

Catalog No. 502707344
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7344 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7344 Supplier Abcam Supplier No. AB12071550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent antidepressant and serotonin-norepinephrine reuptake inhibitor with greater than 99% purity.

  • Molecular Weight: 313.87 g/mol
  • Purity: >99%
  • Form: Solid
  • Function: Serotonin-norepinephrine reuptake inhibitor; antidepressant
  • Binding Affinity: Ki values are 82 nM for SERT and 2480 nM for NET; displays 30-fold higher affinity for SERT than NET in vitro
  • Storage: Store at ambient
  • Shipping: Ambient - Can ship with ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Content And Storage Store at ambient temperature.
Target Molecule SERT, NET
Form Solid
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.