Learn More
Thomas Scientific Vent Filter, 0.22um, Non-Sterile, 25/pack
SDP
Supplier:  Thomas Scientific 1170A25
Encompass
Foxx Life Science Carboy Vent Filters are used for hydrophobic venting and solvent filtration applications. The PTFE filter membrane is ideal for maintaining purified water stored in carboys, or for sterile venting during a slow exhaust/liquid autoclave cycle of EZgrip® carboys. All materials comply with USP Class VI standards, and are autoclavable up to 10 times. Hydrophobic PTFE membrane is reinforced with polypropylene housing and features a stepped hosebarb. Available in Non-sterile 1.00µm, 0.2µm pore sizes with platinum-cured silicone tubing, or in bulk packs (without tubing).
Catalog No. 50-145-8499
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.