Learn More
Eisco Labs Vented Safety Goggles with Adjustable Elastic Strap
Catalog No. 12141693 Shop All Eisco Products
Change view
Click to view available options
Frame Type:
Flexible Temples
Catalog No. Frame Type
12-141-693 Flexible Temples
1 options
Catalog No. 12-141-693 Supplier Eisco Labs Supplier No. G103CLR
  • $5.06
    $4.65 / Each of 1
    Save $0.41
    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Provide reliable eye protection for laboratory environments.
  • Industrial quality safety goggles - Single pair of safety goggles designed to protect your eyes from chemical splashes, high speed particles, dust and UV wavelengths 360-380. Lightweight, durable polycarbonate lens.
  • Direct ventilation - Direct ventilation prevents fogging of the goggle lenses.
  • Comfortable vinyl frame - The soft, flexible vinyl frame and pliable nose bridge provides a comfortable and secure fit. Designed to be able to fit over prescription eyeglasses.
  • Adjustable fit - Goggles feature a freely adjustable, soft elastic head strap for a universal fit. Strap measures 13.5 in. in total length.
  • Great for professional laboratories, classrooms and household - Designed for use in professional laboratories, but are ideal for science classrooms and household use.
  • Industrial quality safety goggles. Single pair.
  • Designed to protect your eyes from chemical splashes, high speed particles, dust and UV wavelengths 360-380.
  • Lightweight, durable polycarbonate lens.
  • The soft, flexible vinyl frame and pliable nose bridge provides a comfortable and secure fit.
  • Direct ventilation prevents fogging of the goggle lenses.
  • Designed to be able to fit over prescription eyeglasses.
  • Goggles feature a freely adjustable, soft elastic head strap.
  • Designed for use in professional laboratories but is ideal for science classrooms and household use.
  • Goggles measure 6 in. wide, 3 in. long and 3 in. tall.
  • Adjustable, head strap elastic measures 13.5 in. in total length and is 0.5 in. thick.
Specifications
Lens Coating UV-treated
Lens Tint Clear
Product Type Safety Goggles
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Vinyl
UV Protection UV380
Frame Type Flexible Temples