Specifications
| Certifications/Compliance | ANSI Z87.1 ,CSA Z94.3-07 |
| Lens Tint | Blue, Mirror |
| Frame Color | Black |
| Weight (Metric) | 27 g |
| Lens Material | Polycarbonate |
| Frame Material | Nylon (6.6) |