Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ VentureGear Tactical™ Stonewall™ Safety Eyewear

Wrap-around, sunglass style

Specifications

Certifications/Compliance ANSI Z87.1,CAN/CSA Z94.3-07
Frame Color Black
Weight (Metric) 28 g
Lens Material Polycarbonate
Frame Material Nylon
View More Specs

Products 2
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
19-156-044
View Documents
Pyramex™
VGSB418T
Bronze N/A N/A N/A
19-156-046
View Documents
Pyramex™
VGSB465T
Blue, Mirror N/A N/A N/A
Description

Description

  • Durable, flexible nylon frame and straight-back temples
  • Built-in rubber nosepiece
  • Anti-fog, scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Passes MIL-PRF 32432 High Velocity Impact Standards
  • Specifications

    Specifications

    ANSI Z87.1,CAN/CSA Z94.3-07
    28 g
    Nylon
    Black
    Polycarbonate
    Videos
    SDS
    Documents

    Documents

    Product Certifications
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.