Learn More
Pyramex™ Venture Gear VG80 Series Ear Muffs
Catalog No. 19156019 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Black graphite pattern
Gray
High-visibility Lime
High-visibility Orange
Pink
Purple
Teal
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-156-019 Purple
19-156-014 Gray
19-156-017 High-visibility Lime
19-555-711 High-visibility Orange
19-156-015 Pink
19-156-018 Teal
19-156-016 Black graphite pattern
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-156-019 Supplier Pyramex™ Supplier No. VGPM8065C
Only null left
Add to Cart
Add to Cart
Venture Gear VG80 series of ear muffs are comfortable and stylish with a variety of colors to choose from.
  • NRR 26 dB increased to 36 dB when worn with ear plugs
  • Low-profile design
  • Height adjustment to fit all sizes
  • Fold-away padded headband
Specifications
Color Purple
ANSI S3.1974 Yes
Cushion Material PVC, Polyurethane
Cushion Color Black
Headband Material POM
Model VGPM8065C
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 26 dB
Headband Color Black
Style Over-the-head
Foldable Yes