Learn More
Labconco Ventus II Canopy
SDP
Catalog No. 13118380 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
8 ft. Logic Biosafety Cabinet
Catalog No. For Use With (Equipment)
13-118-380 8 ft. Logic Biosafety Cabinet
1 options
Catalog No. 13-118-380 Supplier Labconco Supplier No. 3889214
Add to Cart
Add to Cart
Ventus II Canopy kit for 8 ft. Logic Biosafety Cabinets.
  • Kit contains two Ventus II canopies for an 8 ft. Logic Biosafety Cabinet.
  • Airflow alarms detect loss of exhaust within one second and trigger an audible and visual alarm on the line-of-sight touchscreen display.
  • Certifier installation and calibration required.
Specifications
Product Type Exhaust Safety Cabinets Kit
For Use With (Equipment) 8 ft. Logic Biosafety Cabinet