Learn More
Plasma Etch Inc VENUS25 CONFIGURATION 1 MFC
SDP
Supplier:  Plasma Etch Inc PE25VUG1
Encompass
Venus25 Configuration (1 MFC):Includes PC/laptop controller for full PC control with multiple recipe storage, datalogging and 1 Mass Flow Controllers.
Catalog No. NC2865641