Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Verapamil Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607702 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-702 1 g Amber Glass Bottle
18-607-703 5 g Amber Glass Bottle
18-607-704 25 g Amber Glass Bottle
18-607-705 100 g Amber Glass Bottle
18-607-706 1 kg Amber Glass Bottle
18-607-707 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-607-702 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. V10721GM
Add to Cart
Add to Cart

V1072, 152-11-4, C27H38N2O4·HCl

Spectrum™ Chemical Verapamil Hydrochloride, USP belongs to a class of drugs known as calcium channel blockers and is used to treat high blood pressure. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 152-11-4
Molecular Formula C27H39ClN2O4
Molecular Weight (g/mol) 491.07
InChI Key DOQPXTMNIUCOSY-UHFFFAOYNA-N
IUPAC Name hydrogen 2-(3,4-dimethoxyphenyl)-5-{[2-(3,4-dimethoxyphenyl)ethyl](methyl)amino}-2-(propan-2-yl)pentanenitrile chloride
SMILES [H+].[Cl-].COC1=CC=C(CCN(C)CCCC(C#N)(C(C)C)C2=CC=C(OC)C(OC)=C2)C=C1OC

Specifications

CAS Max % 1
pH 4.5 to 6.5
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 1 g
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.