Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Veratramine | 60-70-8 | 99.8% | 409.60 | C27H39NO2 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN083710MG

Encompass_Preferred

Veratramine is a biologically active alkaloid supplied for research use. It modulates SIGMAR1 and inhibits the PI3K/Akt/mTOR signaling pathway, with reported anti-tumor, anti-inflammatory, and neuroprotective activities. The product is offered as a solid standard and as a solution formulation for in vitro and analytical applications.

  • High purity: 99.8% by manufacturer analysis.
  • Molecular weight 409.60 and formula C27H39NO2.
  • CAS number 60-70-8 for unambiguous identification.
  • Available in multiple solid pack sizes, including 10 mg.
  • Also available as a 10 mM solution in DMSO for direct assay use.

Catalog No. 50-002-95687


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.