Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Veratridine, ≥97%

Catalog No. AAJ62324MF
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62324MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62324MF Supplier Thermo Scientific™ Supplier No. J62324MF

Activates sodium ion channels

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) Activates sodium ion channels
Assay ≥97%
CAS 71-62-5
Molecular Formula C36H51NO11
Formula Weight 673.80g/mol
Synonym 3-Veratroylveracevine
Solubility Soluble in ethanol or DMSO; Freely soluble in chloroform; Also soluble in 1M HCl
Product Type Signal Transduction Reagent
Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.