Learn More
MCR Safety Verdict Goggle
MCR Safety Verdict Goggle features a body design of the that provides a wide, unobstructed field of vision and can be worn with most half-mask respirators.
$11.24
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
18-999-2611
|
MCR Safety
2410 |
Not foam lined |
Each for $11.24
Pack of 12 EA for $134.90
|
|
|||||
18-999-4574
|
MCR Safety
2410F |
Foam-lined | N/A | N/A | N/A | ||||
Description
- Protective PVC body permits a comfortable fit over prescription eyewear
- Optional soft foam lining cushions the face and improves the seal
- Offered in both scratch-resistant polycarbonate lens and anti-fog lens
- Available with either a wide elastic headband or a neoprene headband
- Meets or exceeds ANSI Z87+ and CSA Z94.3.
Specifications
Clear | |
Smoke | |
Indirect vent |