Learn More
Medtox™ Diagnostics VERDICT™-II Buprenorphine
Description
- 510(k) cleared
- Screens for buprenorphine
- Quality management system conforms with ISO 13485
- Accurate screening results in minutes
- Made in the USA
Specifications
Specifications
| CE Marker | No |
| Clia Complexity | Moderate |
| Detectable Analytes | Buprenorphine (BUP) |
| Disposable | Single-use |
| For Use With (Application) | Drug Panel (Urine) |