Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Verdinexor
GREENER_CHOICE

Catalog No. p-7250112
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-691-0 10 mg
75-695-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 75-691-0 Supplier Tocris Bioscience™ Supplier No. 7569/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective exportin-1 (XPO1/CRM1) inhibitor

Verdinexor is a selective exportin-1 (XPO1/CRM1) inhibitor. It potently inhibits influenza virus ribonucleoprotein (vRNP) export by disrupting XPO1-NEP binding and the replication of influenza virus A and B strains in vitro, including H1N1, H5N1 and H7N9 (IC50 values in the range 40 - 420 nM range). Verdinexor limits influenza A virus spread in mouse lung in vivo. In neuroblastoma, verdinexor suppresses cell proliferation and induces cell apoptosis. It also induces P53 nuclear accumulation and induces G0/G1 phase cell cycle arrest by activating P53 function. Verdinexor inhibits tumor growth in in vivo mouse models. Orally bioavailable.

Specifications

Chemical Name or Material (2 Z)-3-[3-[3,5-Bis(trifluoromethyl)phenyl]-1 H-1,2,4-triazol-1-yl]-2-propenoic acid 2-(2-pyridinyl)hydrazide
CAS 1392136-43-4
Quantity 10 mg
Target Antiviral Compounds
Molecular Formula C18H12F6N6O
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.