Learn More
Apexbio Technology LLC SP2509 1423715-09-6 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B48945.1
Encompass_Preferred
SP2509 (CAS 1423715-09-6) is a small molecule antagonist of lysine-specific demethylase 1 (LSD1/KDM1A) a histone-modifying enzyme responsible for demethylating mono- and dimethylated histone H3 lysine 4 (H3K4) SP2509 potently inhibits LSD1 activity (IC50 13 nM) without affecting monoamine oxidase isoforms MAO-A or MAO-B In acute myeloid leukemia (AML) cells SP2509 suppresses LSD1 function reduces colony formation induces apoptosis enhances promoter-specific H3K4 trimethylation and stimulates differentiation through upregulation of p53 p21 and C/EBP Its efficacy has been demonstrated in AML xenograft models supporting its utility as a tool compound for cancer research
Catalog No. 50-000-63079
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.