Learn More
PBL Assay Science VeriKine Human Interferon Gamma Receptor 1 ELISA Kit
Supplier:  PBL Assay Science 415801
Encompass_Preferred
Accurate quantitation of soluble and membrane IFN-Gamma Receptor 1
Catalog No. NC2047817
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.