Learn More
Honeywell™ VeriPRO Fit-Testing System
Catalog No. 19085596 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Packaging Type:
Carrying Case
Catalog No. Packaging Type
19-085-596 Carrying Case
1 options
Catalog No. 19-085-596 Supplier Honeywell™ Supplier No. VERIPRO
Add to Cart
Add to Cart
Request a Quote
VeriPRO’s three-part process checks the effectiveness of an employee’s ear plug fit in each ear over a range of frequencies. This information is then captured in individual reports, accessible by the safety manager.
  • Measures the Personal Attenuation Rating (PAR) for each employee
  • Works with any ear plug, corded or uncorded, from any manufacturer
  • Fulfills regulatory requirements to ensure proper initial fitting of hearing protection
  • Documents training and proper fit for new or existing employees
  • Provides an ideal opportunity for one-on-one education
  • Minimum Requirements 700 Mhz CPU, 256 MB RAM
  • Software Requirements Windows 7 or Windows 10, Microsoft SQL Server Express, Microsoft .NET Framework 4.5
  • Rating Systems - NRR (North America), SNR (Europe/Asia), SLC80 (Australia)
  • Hard Disk Requirements 800 MB
Specifications
For Use With (Equipment) Howard Leight™ VP Headphones
Model VeriPRO™
Packaging Type Carrying Case