Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ VeriShield™ 100 Series Behind-the-Neck Ear Muffs

Catalog No. 19085532 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
CSA Z94.2:
Class AL
Class B
Noise Reduction Rating (NRR):
22 dB
25 dB
28 dB
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. CSA Z94.2 Noise Reduction Rating (NRR)
19-085-532 Class B 22 dB
19-085-534 Class AL 25 dB
19-085-535 Class AL 28 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-085-532 Supplier Honeywell™ Supplier No. 1035112VS
Add to Cart
Add to Cart

VeriShield™ Behind-the-Neck Ear Muffs fit as a neckband to complement any other PPE or equipment the worker may be wearing or using.

  • Fits around the neck from behind
  • Wide cup opening is ideal for workers with large ears, or who wear hearing aids
  • Lightweight cushioned headband
  • Memory foam ear cushions
  • Oil-resistant cup surface withstands heavy-work environments and frequent cleaning
  • Features Sound Management Technology (SMT) which improves employee safety by blocking harmful noise while allowing voices and signals to be heard more naturally
  • Designed for use with other PPE

Specifications

Color Black
ANSI S3.1974 Yes
CSA Z94.2 Class B
Cushion Color Black
For Use With (Application) Construction, Manufacturing, Oil and Gas, Mining, Steel and Metals, Welding, Agriculture
Model VS110N
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 22 dB
Product Type VeriShield
Dielectric No
Material ABS
Style Behind-the-head
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.