Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ VeriShield™ 100 Series Over-the-Head Ear Muffs

Catalog No. 19085524 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
CSA Z94.2:
Class A
Class AL
Color:
Black
High-visibility Yellow
Foldable:
No
Yes
Noise Reduction Rating (NRR):
24 dB
26 dB
27 dB
30 dB
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. CSA Z94.2 Color Foldable Noise Reduction Rating (NRR)
19-085-524 Class A Black No 24 dB
19-085-526 Class A Black Yes 24 dB
19-085-477 Class A Black No 26 dB
19-085-529 Class AL High-visibility Yellow Yes 27 dB
19-085-482 Class AL Black No 30 dB
19-085-531 Class AL High-visibility Yellow No 30 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-085-524 Supplier Honeywell™ Supplier No. 1035100VS
Add to Cart
Add to Cart

VeriShield™ Over-the-Head style ear muffs feature air flow control technology for maximum attenuation without increasing pressure on the head.

  • Comfortably fits a wide range of head sizes
  • Wide cup opening is ideal for workers with large ears, or who wear hearing aids
  • Lightweight cushioned headband
  • Memory foam ear cushions
  • Oil-resistant cup surface withstands heavy-work environments and frequent cleaning
  • Features Sound Management Technology (SMT) which improves employee safety by blocking harmful noise while allowing voices and signals to be heard more naturally
  • Designed for use with other PPE

Specifications

Color Black
ANSI S3.1974 Yes
CSA Z94.2 Class A
Cushion Color Black
For Use With (Application) Construction, Manufacturing, Oil and Gas, Mining, Steel and Metals, Welding, Agriculture
High Visibility No
Model VS110
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 24 dB
Product Type VeriShield
Dielectric No
Material ABS
Style Over-the-head
Foldable No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.