Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Beryllium (Be) Standard for AAS 1000 ppm in 2% HNO3/1% HF, Ricca Chemical
SDP

Catalog No. RV010067500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010067500 500 mL
RV010067100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010067500 Supplier Ricca Chemical Company Supplier No. RV010067500N
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 2% HNO3/1% HF
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 13597-99-4 , 7697-37-2 , 7664-39-3 , 7732-18-5
Molecular Formula BeN2O6
Molecular Weight (g/mol) 133.02
InChI Key RFVVBBUVWAIIBT-UHFFFAOYSA-N
PubChem CID 26126
IUPAC Name beryllium;dinitrate
SMILES [Be+2].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-]

Specifications

Packaging Natural Poly Bottle
CAS Min % 93.61%, 1.96%, 1.45%, 0.98%
CAS Max % 97.43%, 2.04%, 1.51%, 1.02%
Specific Gravity 1.01
Physical Form Liquid
UN Number UN3287
Melting Point Approximately 0°C
Type Single-element Standard Solution
Quantity 500 mL
Grade High Purity
Chemical Name or Material Beryllium
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.