Learn More
Vernier Chemical Polarimeter Accessory Cells
SDPSCI_ED
Catalog No. S16118ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Vernier Chemical Polarimeter
Catalog No. For Use With (Equipment)
S16118ND Vernier Chemical Polarimeter
1 options
Catalog No. S16118ND Supplier Vernier Supplier No. CELLSPOL
Add to Cart
Add to Cart
Reduces inconsistencies between runs.
  • Package of four replacement sample cells for the Vernier Chemical Polarimeter
  • Fit snugly in Chemical Polarimeter to reduce inconsistencies between runs
  • Easy-pour spout for transferring solutions safely and efficiently
  • Flat-bottom glass tube
  • Dimensions: 20mm inner dia.x 150mm tall
  • Screen-printed ruler with cm markings
Specifications
Type Replacement Sample Cell
Material Glass
For Use With (Equipment) Vernier Chemical Polarimeter
Dimensions (Dia. x H) 20 x 150 mm