Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier KidWind Balsa Blade Sheets
SDPSCI_ED

Catalog No. S04802ND Shop All Vernier Software Products
Change view
Click to view available options
Quantity:
10 Pk.
100 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04802ND 10 Pk.
S04801ND 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04802ND Supplier Vernier Supplier No. KWBBS10
Add to Cart
Add to Cart

Lightweight and stiff, suitable for making experimental wind turbine blades.

  • Recommended for college, high school, and middle school science
  • Easy to cut, carve, and shape
  • Used as wood skeletons inside fiberglass for real wind turbine blades
  • L x W: 30.48 x 7.6cm
  • Thickness: 2.5mm

Specifications

Product Type Balsa Blade Sheets
Material Balsa Wood
For Use With (Application) Making experimental wind turbine blades
Quantity 10 Pk.
Thickness 2.35 mm
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.