Learn More
Vernier KidWind simpleGEN Easy-to-build AC Generator Kit
SDPSCI_ED
Catalog No. S04824ND Shop All Vernier Software Products
Change view
Click to view available options
Product Type:
simpleGEN AC Generator Kit
simpleGEN AC Generator Kit, Classroom Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S04824ND simpleGEN AC Generator Kit
S04825ND simpleGEN AC Generator Kit, Classroom Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04824ND Supplier Vernier Supplier No. KWSGEN
Add to Cart
Add to Cart
Includes
Wind Turbine Hub, Alligator clip cord pair, Digital multimeter, Sandpaper 1/8 sheet, Magnet holder, Ceramic magnet, 8 in. hex shaft with hub quick connect, 4 in. hex shaft, Simple hand generator housing set, Dowels, Red superbright LED, Bi-polar LED bulb and 28 gauge magnet wire with spool
Allows students to explore the basics of electrical generator design.
  • To demonstrate Faraday's law, light LEDs
  • To perform experiments on how coils, magnets, and rotation affect electrical generation
  • Can be converted to simple motor by simple modifications
  • Also available in classroom pack which includes materials to build 10 generators and to convert two generators to wind turbines
  • Recommended for College, High School, and Middle School Science
Activities Performed:
  • Generate electricity
  • AC power output and measures output voltage
  • Compare different designs (coils, magnets, spinning)
Concepts:
  • Understanding clean energy
  • Exploring energy transformations, relationship between energy and forces
  • Collecting, analyzing and interpreting data with advanced tools
  • Designing controlled experiments
Warranty and Services
5-year limited warranty (excludes consumables)
Specifications
For Use With (Application) Allows students to explore the basics of electrical generator design, demonstrate Faraday's law, light LEDs and how coils, magnets, and rotation affect electrical generation
Product Type simpleGEN AC Generator Kit
WARNING: Cancer - www.P65Warnings.ca.gov