Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Motion Detector For CBL System
SDPSCI_ED

Catalog No. S405904 Shop All Vernier Software Products
Change view
Click to view available options
Range:
0.15 to 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
S405904 0.15 to 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S405904 Supplier Vernier Supplier No. MDBTD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Motion detector to LabQuest/LabQuest mini/LabPro/CBL™ cable, user manual

Used to measure position, velocity and acceleration of moving objects.

  • Sensitivity switch reduces noise
  • Pivoting head for flexibility
  • Range: 0.15 to 6m
  • Resolution: 1mm

Warranty and Services

5 years

Specifications

Type Motion Detector
Includes Motion detector to LabQuest/LabQuest mini/LabPro/CBL™ cable, user manual
Warranty 5 Years
For Use With (Application) Used to measure position, velocity and acceleration of moving objects
Range 0.15 to 6
Resolution 1mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.