Learn More
Vernier ProScope HR Digital Microscope Kits
SDPSCI_ED
Catalog No. S16091ND Shop All Vernier Software Products
Change view
Click to view available options
Description:
Biology ProScope HR Kit
ProScope HR Deluxe Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
S16091ND ProScope HR Deluxe Kit
S16093ND Biology ProScope HR Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S16091ND Supplier Vernier Supplier No. BDDLX
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
High-resolution handheld digital microscope for capturing images, videos, and time-lapse photographs.
The ProScope HR is a USB handheld digital microscope designed for use with computer. With a high-quality CMOS sensor and universal lens mount, the ProScope HR is a powerful addition to any classroom.
Biology Kit:
Use ProScope HR as dissecting microscope or attach ProScope HR to regular microscope for even more magnification.
Includes:
  • ProScope HR base
  • ProScope HR software
  • 50X lens
  • 1-10X lens
  • C-mount adapter with IR cut filter
  • Microscope lens tube adapter
  • Camera stand
  • Rugged hard-side case
Deluxe Kit:
For maximum magnification with or without regular microscope, deluxe kit has covered with its lens ensemble and accessories.
Includes:
  • ProScope HR base
  • ProScope HR software
  • 50X lens
  • 1-10X lens
  • 100X lens
  • 200X lens
  • C-mount adapter with IR cut filter
  • Microscope lens tube adapter
  • Camera stand
  • Rugged hard-side case
Proscope HR Specifications:
  • Image sensor: 1/4 in. high resolution 1.3 megapixel sensor
  • Connection type: USB
  • Image capture resolutions:
  • 320x240 pixels
  • 640x480 pixels
  • 1280x1024 pixels
  • Video capture resolutions:
  • 320x240 pixels at @30 fps
  • 640x480 pixels at @15 fps
  • 1280x1024 pixels at @3.75 fps
Specifications
Description ProScope HR Deluxe Kit
Includes Bodelin Proscope HR, three additional lenses and accessories
Required Accessories Windows: Windows™ XP service pack 2 or newer, Quicktime 7.0 or newer, DirectX 9 or newer, available USB 2 port, Pentium 4 or equivalent; Macintosh: Mac OS 10.4.8 or newer, Quicktime 7.0 or newer, factory-installed USB 2 port, G4, G5, or Intel processor; Chromebook: Camera app and available USB 2 port