Learn More
Sigma Aldrich Fine Chemicals Biosciences Metal drum pump fits 5gal.1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5553631EA
Encompass
High quality fM approved pump with grounding wires and clamps. for dispensing solvents and non-corrosive liquids. Pump includes adapter Z562254 for mounting pump to the Sigma-Aldrich 18L-CS or 20 L flex-spout can plus adapters for can threads 1 3/4 inch 2 1/8 inch 2 1/4 inch and 2 7/8 " Dispenses 100 mL/stoke.
Catalog No. 50-290-5696
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.