Learn More
3M™ Versaflo™ Multi Layer Peel-Off Visor Cover
Catalog No. 17000660 Shop All 3M Products
This item is not returnable. View return policy
Catalog No. 17-000-660 Supplier 3M™ Supplier No. 7010385074
Add to Cart
Add to Cart
This item is not returnable. View return policy
Versaflo™ multi layer peel-off visor cover is a multi-layer transparent covering for S-Series hood assemblies to protect the visor from paint overspray, contaminants, and scratches.
  • 4 layer peel off film helps protect the visor from contaminants and scratches
  • Multi-layer peel-off helps extend the life of the hood in a paint spray application
  • Full perimeter adhesive enables for easy attachment to visor
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Material Peel-Off
For Use With (Application) Painting, Spraying
For Use With (Equipment) 3 M™ S-Series Headgear