Description
- Provide extended use of powered air purifying respirator systems
- 3M Catalog# GVP-110, GVP-210, TR-341N, TR-344N, TR-641N, TR-644N, TR-941N, TR-944N, 520-04-24
Compliance
NIOSH approved.
Specifications
Specifications
| Product Type | Single-Unit Charging Station |
| For Use With (Equipment) | Versaflo TR-300 PAPRs |
| No. of Ports | One |