Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ PAPR System Filters and Cartridges

Catalog No. 19268010 Shop All 3M Products
Change view
Click to view available options
Product Type:
Cartridge
HE Filter
Filter Type:
Combination HE Particulate/Acid Gas/HEPA
Combination HE Particulate/Ammonia/Methylamine
Combination HE Particulate/Formaldehyde
Combination HE Particulate/HEPA and Nuisance Organic Vapor/Acid Gas
Combination HE Particulate/Organic Vapor/Acid Gas/HEPA
Combination HE Particulate/Organic Vapor/HEPA
Particulate
Color:
Black/Magenta
Magenta
Magenta/Green
Magenta/Olive
Magenta/White
Magenta/Yellow
For Use With (Application):
Healthcare, Pharmaceutical, Heavy Industrial, Manufacturing, General Manufacturing
Pharmaceutical
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Filter Type Color For Use With (Application)
19-268-010 Cartridge Combination HE Particulate/Organic Vapor/HEPA Black/Magenta Pharmaceutical
19-268-007 HE Filter Particulate Magenta Healthcare, Pharmaceutical, Heavy Industrial, Manufacturing, General Manufacturing
19-268-012 Cartridge Combination HE Particulate/Acid Gas/HEPA Magenta/White Pharmaceutical
19-268-011 Cartridge Combination HE Particulate/Organic Vapor/Acid Gas/HEPA Magenta/Yellow Pharmaceutical
19-268-013 Cartridge Combination HE Particulate/Ammonia/Methylamine Magenta/Green Pharmaceutical
19-268-014 Cartridge Combination HE Particulate/Formaldehyde Magenta/Olive Pharmaceutical
19-268-009 Cartridge Combination HE Particulate/HEPA and Nuisance Organic Vapor/Acid Gas Magenta Healthcare, Pharmaceutical, Heavy Industrial, Manufacturing, General Manufacturing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-268-010 Supplier 3M™ Supplier No. 7100021253
Add to Cart
Add to Cart

Compatible with powered air purifying respirators in the TR-600 Series

  • Filters and cartridges work with belt-mounted Versaflo PAPR units
  • Recommended for use in healthcare, pharmaceutical, heavy industrial, manufacturing and/or general manufacturing environments
  • NIOSH approved
  • 3M Catalog# TR-6320N, TR-6350N, TR-6360N, TR-6510N, TR-6530N, TR-6710N-5, TR-6820N

Compliance

NIOSH approved.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH
Color Black/Magenta
Filter Type Combination HE Particulate/Organic Vapor/HEPA
Product Type Cartridge
For Use With (Application) Pharmaceutical
For Use With (Equipment) TR-600 PAPR
Model TR-6510N
Quantity 5/Case
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.