Learn More
Description
- Filters and cartridges work with belt-mounted Versaflo PAPR units
- Recommended for use in healthcare, pharmaceutical, heavy industrial, manufacturing and/or general manufacturing environments
- NIOSH approved
- 3M Catalog# TR-6320N, TR-6350N, TR-6360N, TR-6510N, TR-6530N, TR-6710N-5, TR-6820N
Compliance
NIOSH approved.
Specifications
Specifications
| Certifications/Compliance | NIOSH |
| Color | Magenta/White |
| Filter Type | Combination HE Particulate/Acid Gas/HEPA |
| Product Type | Cartridge |
| For Use With (Application) | Pharmaceutical |
| For Use With (Equipment) | TR-600 PAPR |
| Model | TR-6320N |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.