Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ Respiratory M-Series Helmet Assembly with Leather Shroud

Catalog No. 19070937 Shop All 3M Products
Change view
Click to view available options
Size:
Standard
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-070-937 Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-937 Supplier 3M™ Supplier No. 7100168650
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Versaflo™ Respiratory M-Series Helmet Assembly with Leather Shroud includes a flame resistant helmet cover and Speedglas™ Weld Shield.

  • New, more durable leather shroud
  • 10% weight reduction when compared to L-905SG
  • Protection from welding arc (ANSI Z87) plus spark and splatter
  • Larger viewing area compared to L-905SG
  • Able to lift visors in non-hazardous areas to help facilitate communications
  • Head top can be used for most applications including arc gouging, plasma cutting, grinding, MIG, TIG, and GMAW welding
  • When used in conjunction with NIOSH approved air source, is designed to help protect the user from certain airborne contaminants, and certain head, eye, face, and welding hazards
  • Compatible with 3M™ Adflo, 3M™ Versaflo™ Powered Air Purifying Respirators, and 3M™ Supplied Air
  • Compatible with 9100XXi ADF with Natural Color Technology
  • 3M Catalog# M-409SG

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI Z89.1 Type I, Class G
Color Black
For Use With (Application) Welding
Includes 3M™ Versaflo™ Respiratory M-Series Helmet Assembly Leather Shroud, Nomex™ Headcover, Speedglas™ Weld Shield
Material Leather
Model M-409SG
Product Type Headgear
Quantity 1 Ea.
Size Standard
Visor Type Clear
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.