Learn More
Cytiva (Formerly Pall Lab) Versapor Acrylic Copolymer 1200 Membrane Disc Filter, 1.2 m - 25 mm (100/pkg)
Catalog No. NC1703874 Shop All Cytiva Products
Encompass
Ideal for prefiltration of difficult-to-filter solutions and serum.-For prefiltration-Excellent retention-Acrylic copolymerDesigned to be used in applications requiring excellent retention properties combined with tensile strengths in excess of 200 bar (3000 psi).Filter membrane is made of acrylic copolymer on a nonwoven support. Strong, flexible, and nonshedding, both dry and wet. Delivers high flow rates at low differential pressures. Can be used as a prefilter to replace glass fiber filters. UV or gamma sterilizable. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1703874 Supplier Cytiva Supplier No. 66393
Ideal for prefiltration of difficult-to-filter solutions and serum.-For prefiltration-Excellent retention-Acrylic copolymerDesigned to be used in applications requiring excellent retention properties combined with tensile strengths in excess of 200 bar (3000 psi).Filter membrane is made of acrylic copolymer on a nonwoven support. Strong, flexible, and nonshedding, both dry and wet. Delivers high flow rates at low differential pressures. Can be used as a prefilter to replace glass fiber filters. UV or gamma sterilizable. This product is not approved for use in medical, clinical, surgical or other patient applications.