Learn More
*LETCO MEDICAL Versatile™ Cream Base 500g
Supplier: *LETCO MEDICAL 501014
Versatile™ Cream Base is a highly elegant vanishing cream. It offers versatility for compounding challenges as it retains its consistency with a broad variety of APIs and solvents. Versatile™ Cream Base offers a sophisticated, elegant skin feel, helping to improve patient’s adherence. Versatile™ Cream Base is ideal for both prescription compounding and basic skin care and is often used in patients with vulnerable, sensitive, balanced and (very) dehydrated skin types. **Requires a copy of state medical license that has address matching the ship to, for every purchase order**