Learn More
Ansell™ VersaTouch™ Protective Sleeve
Catalog No. 19821921 Shop All Ansell Products
Change view
Click to view available options
:
21 in.
Catalog No.
19-821-921 21 in.
1 options
This item is not returnable. View return policy
Catalog No. 19-821-921 Supplier Ansell™ Supplier No. 284301
This item is not returnable. View return policy
Ansell™ VersaTouch™ Protective Sleeve features an engineered yarn containing steel and glass fibers combined with Dyneema™ to provide ANSI Level 4 cut resistance in food applications that present risks to a worker’s arms
  • Sleeve allows for more mobility than other cut protective sleeves, leading to increased productivity and yield
  • Pulls on easily and clips into place for a relaxed fit and also features Tuff Cuff II Technology.
  • Breakthrough in cut protection for the Food Processing industry
  • High-level cut resistant
  • Helps retain cuff shape, keeping gloves secure and reducing worker injury
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum, Plastic