Learn More
StatLab Versenate Decal Solution, 1 Liter
SDP
Supplier:  StatLab DCVERLT
Encompass_Preferred
A bone decalcifier that requires extensive use of special stain techniques. Ideal for iron stains. This decal can take several days to complete the decalcification process.
Catalog No. NC9445371
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.