Learn More
Description
1 μg/mL of MA5-27638 was sufficient for detection of Versican in 20 μg of mouse brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects >350kDa. This antibody was formerly sold as clone S351-23.
Specifications
Specifications
| Antigen | Versican |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | N351/23 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.4 |
| Gene | VCAN |
| Gene Accession No. | P13611, Q62059, Q9ERB4 |
| Gene Alias | 5430420N07Rik; 9430051N09; Chondroitin sulfate proteoglycan 2; chondroitin sulfate proteoglycan 2 (versican); chondroitin sulfate proteoglycan core protein 2; Cspg2; DPEAAE; ERVR; GHAP; glial hyaluronate-binding protein; hdf; heart defect; large fibroblast proteoglycan; NG2; PGM; PG-M; PG-M core protein; PG-M(V0); PG-M(V1); V1 Neo; VCAN; versican; versican core protein; Versican core protein precursor (Large fibroblast proteoglycan) (Chondroitin sulfate proteoglycan core protein 2) (PG-M); versican proteoglycan; Versican V0; WGN; WGN 1; WGN1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
