Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Verso 1-Step RT-PCR Kit ReddyMix, with ThermoPrime Taq

Catalog No. AB1454LDB Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1454LDB 200 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1454LDB Supplier Thermo Scientific™ Supplier No. AB1454LDB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific Verso 1-Step RT-PCR Kit ReddyMix with ThermoPrime Taq is a complete set of RT-PCR reagents that combines everything needed for RT-PCR.

Thermo Scientific Verso 1-Step RT-PCR Kit ReddyMix with ThermoPrime Taq is a complete set of RT-PCR reagents that combines everything needed for RT-PCR. The Verso RT enzyme delivers high processivity and can be used at temperatures up to 57°C, delivering effective transcription through difficult RNA secondary structures. Verso RT also shows a better dynamic range than standard AMV or M-MuLV enzymes, with increases in template concentration giving rise to corresponding increases in product generated over a wider range of starting material concentrations.

An optional RT enhancer is provided with the kit and can be used during the RT step to remove any DNA contamination, eliminating the need for DNase I treatment.

This Verso 1-step RT-PCR system comes with the ReddyMix option, where the master mix contains a dye and precipitant that allow RT-PCR reactions to be loaded directly onto an electrophoresis gel. The red dye migrates between bromophenol blue and xylene cyanol at approximately 300 bp, depending on agarose concentration.

Features of the Verso 1-Step RT-PCR Kit ReddyMix with ThermoPrime Taq include:
• ReddyMix formulation—eliminates post-PCR steps required for electrophoresis
• Eliminates the need for DNase treatments—RT Enhancer (optional) prevents gDNA carry-over
• RT and PCR steps performed in single sealed tube. providing convenience and reduced contamination risk
• Verso RT enzyme is highly sensitive with a broad dynamic range—accurately detect RNA input from 1 pg to 1 μg
• Wide working temperature range (42–57°C) for success with GC-rich and other difficult templates
• RNase Inhibitor included in RT enzyme mix

Applications
• Gene expression analysis
• Viral/bacterial detection
• 1-step RT-PCR
• High throughput applications

Related products
Verso 1-Step RT-PCR Kit ReddyMix, with ThermoPrime Taq (Cat. No. AB1454LDA)
Verso 1-Step RT-PCR Kit, with Thermo-Start Taq (Hot Start) (Cat. No. AB1455A)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

• Verso Enzyme Mix
• 2X Master Mix
• RT Enhancer

Store at –20°C.

Detection Method Gel Electrophoresis
Format Kit
PCR Method 1-step RT-PCR
Polymerase ThermoPrime Taq DNA Polymerase
Technique Reverse Transcription
Optimal Reaction Temperature 42°C to 57°C
Quantity 200 reactions
Reverse Transcriptase Verso
Ribonuclease H Activity Yes
Shipping Condition Dry Ice
For Use With (Application) RT-PCR
Final Product Type PCR Amplified cDNA
No. of Reactions 200 Reactions
Reaction Format Premixed Components
Reagent Type Reverse Transcription
Size (Final Product) Up to 11 kb
Starting Material RNA
Show More Show Less
What is the RT Enhancer that is included in all Verso kits?

The RT Enhancer is a unique enzyme that degrades contaminating genomic DNA at the start of the RT step. It is equally as effective as DNase I treatment but more convenient as it eliminates the need for a separate DNase I incubation prior to RT-PCR.

Do I need to treat my RNA with DNase I before running RT-qPCR with Verso cDNA Synthesis Kit?

No, you do not need to treat your RNA with DNase I before running RT-qPCR with the Verso cDNA Synthesis Kit. The RT enhancer included in the Verso kit degrades any contaminating double-stranded DNA during the reverse transcription process, eliminating the need for a separate DNase I treatment.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.