Learn More
Market Lab Vertical Wall Mounted Infant Blood Draw Station
Supplier:  Market Lab 15934WD
Encompass_Preferred
The Vertical Wall Mounted Infant Blood Draw Station by MarketLab is designed for drawing blood from infants and small children in a clinical setting. It features a padded infant platform that adjusts to multiple angles. The station is made with durable steel construction with a powder coated finish. It can be mounted to a wall to save space. The station includes side rails and a restraint strap to safely secure the infant during the procedure. This product provides an ergonomic workspace for phlebotomists to improve workflow and patient care.
Catalog No. NC2749557
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.