Learn More
Scientific Industries VERTICAL 50ML TUBE HOLDER
Supplier:  Scientific Industries SIV506
Encompass_Preferred
Tube Holder holds six 50mL tubes vertically. Holder snaps on to any Vortex-Genie 2 Mixer and the Vortex-Genie Pulse. No additional attachment necessary.
Catalog No. 50-728-071
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.