Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Vertical Freezer Half Rack, Holds (4) 2" Boxes, Rack Dimensions 21 15/16H x 2 1/4D x 5 9/16W" (558 x 57 x 141mm) Compartment Dims 5 3/8H x 2 1/8D x 5 1/4W" (137 x 54.4 x 133mm ), With Locking Rod
Supplier:  CRYSTAL TECHNOLOGY AND INDUSTRIES LR42
Encompass_Preferred
Rack Designed to hold (4) Standard 2"Cardboard Boxes, Plastic (Polypropylene or Polycarbonate) or Metal (Stainless Steel or Aluminum) Vertically, made of corrosion resistant stainless steel, for use in Freezers or LN2 Tanks, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available.
Catalog No. NC2208079
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.