Learn More
Eisco™ Vertical Splash Heads - Pear Shaped
SCI_ED
Catalog No. S29486 Shop All Eisco Products
Change view
Click to view available options
Standard Taper Joint:
To fit flask- 24/29, To fit condenser- 19/26
Catalog No. Standard Taper Joint
S29486 To fit flask- 24/29, To fit condenser- 19/26
1 options
Catalog No. S29486 Supplier Eisco™ Supplier No. CH0854B
Add to Cart
Add to Cart
For use in distillation assemblies with vertical condensers. A variety of flask and condenser cone sizes are available.
  • Vertical splash head
  • Pear shaped trap
  • For use with vertical condensers
  • For use between flask and condenser to prevent carry over of raw liquids
  • Made from high quality borosilicate 3.3 glass
  • Autoclavable
Specifications
For Use With (Application) Distillation
Type Vertical
Material Borosilicate Glass 3.3
Quantity 1
Standard Taper Joint To fit flask- 24/29, To fit condenser- 19/26
1 year limited manufacturer warranty